missing translation for 'onlineSavingsMsg'
Learn More

5-Acetylthiophene-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. A28661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28661G 1 g
A28665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28661G Supplier TCI America Supplier No. A28661G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4066-41-5
Molecular Formula C7H6O3S
Molecular Weight (g/mol) 170.182
MDL Number MFCD00055512
InChI Key LIKIMWYKJUFVJP-UHFFFAOYSA-N
Synonym 5-acetyl-2-thiophenecarboxylic acid, 5-acetyltiophene-2-carboxylic acid, 2-thiophenecarboxylic acid, 5-acetyl, 5-acetyl-2-thenoic acid, acmc-1ae0g, 5-carboxy-2-acetylthiophene, 5-acetylthiophene-2-carboxylicacid, 5-acetyl-thiophen-2-carboxylic acid, 5-acetyl thiophene-2-carboxylic acid, 5-acetyl-2-thiophene-carboxylic acid
PubChem CID 3742727
IUPAC Name 5-acetylthiophene-2-carboxylic acid
SMILES CC(=O)C1=CC=C(S1)C(=O)O

Specifications

Melting Point 211°C
Color Yellow
Quantity 1 g
Formula Weight 170.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Acetylthiophene-2-carboxylic Acid