accessibility menu, dialog, popup

UserName

H3COSOO

CAS RN 4066-41-5

IUPAC Name: 5-acetylthiophene-2-carboxylate
Synonyms: 5-acetylthiophene-2-carboxylate
Molecular Weight (g/mol): 169.17
Molecular Formula: C7H5O3S
InChi Key: LIKIMWYKJUFVJP-UHFFFAOYSA-M
SMILES: CC(=O)C1=CC=C(S1)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)