missing translation for 'onlineSavingsMsg'
Learn More

5-Acetylthiophene-2-carboxylic acid, 98+%

Catalog No. AAL1358814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1358814 25 g
AAL1358806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1358814 Supplier Thermo Scientific Chemicals Supplier No. L1358814

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4066-41-5 | C7H6O3S | 170.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4066-41-5
Molecular Formula C7H6O3S
Molecular Weight (g/mol) 170.182
MDL Number MFCD00055512
InChI Key LIKIMWYKJUFVJP-UHFFFAOYSA-N
Synonym 5-acetyl-2-thiophenecarboxylic acid, 5-acetyltiophene-2-carboxylic acid, 2-thiophenecarboxylic acid, 5-acetyl, 5-acetyl-2-thenoic acid, acmc-1ae0g, 5-carboxy-2-acetylthiophene, 5-acetylthiophene-2-carboxylicacid, 5-acetyl-thiophen-2-carboxylic acid, 5-acetyl thiophene-2-carboxylic acid, 5-acetyl-2-thiophene-carboxylic acid
PubChem CID 3742727
IUPAC Name 5-acetylthiophene-2-carboxylic acid
SMILES CC(=O)C1=CC=C(S1)C(=O)O

Specifications

Melting Point 208°C to 212°C
Quantity 25 g
Beilstein 128897
Formula Weight 170.19
Percent Purity ≥98%
Chemical Name or Material 5-Acetylthiophene-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only