missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Acetylindan 98.0+%, TCI America™
$211.77 - $710.39
Chemical Identifiers
| CAS | 4228-10-8 |
|---|---|
| Molecular Formula | C11H12O |
| Molecular Weight (g/mol) | 160.22 |
| MDL Number | MFCD00021246 |
| InChI Key | HCMWPTFPUCPKQM-UHFFFAOYSA-N |
| Synonym | 5-acetylindane, 5-acetylindan, 1-2,3-dihydro-1h-inden-5-yl ethanone, 1-2,3-dihydro-1h-inden-5-yl ethan-1-one, ethanone,3-dihydro-1h-inden-5-yl, indan-5-ylethan-1-one, 1-indane-5-yl-ethanone, acmc-1arc8, 1-2,3-dihydro-1h-inden-6-yl ethanone, 1-2,3-dihydro-1h-inden-5-yl-ethanone |
| PubChem CID | 233976 |
| IUPAC Name | 1-(2,3-dihydro-1H-inden-5-yl)ethanone |
| SMILES | CC(=O)C1=CC2=C(CCC2)C=C1 |
Chemical Identifiers
| 4228-10-8 | |
| 160.22 | |
| HCMWPTFPUCPKQM-UHFFFAOYSA-N | |
| 233976 | |
| CC(=O)C1=CC2=C(CCC2)C=C1 |
| C11H12O | |
| MFCD00021246 | |
| 5-acetylindane, 5-acetylindan, 1-2,3-dihydro-1h-inden-5-yl ethanone, 1-2,3-dihydro-1h-inden-5-yl ethan-1-one, ethanone,3-dihydro-1h-inden-5-yl, indan-5-ylethan-1-one, 1-indane-5-yl-ethanone, acmc-1arc8, 1-2,3-dihydro-1h-inden-6-yl ethanone, 1-2,3-dihydro-1h-inden-5-yl-ethanone | |
| 1-(2,3-dihydro-1H-inden-5-yl)ethanone |
Specifications
| 4228-10-8 | |
| 85°C | |
| MFCD00021246 | |
| 5-acetylindane, 5-acetylindan, 1-2,3-dihydro-1h-inden-5-yl ethanone, 1-2,3-dihydro-1h-inden-5-yl ethan-1-one, ethanone,3-dihydro-1h-inden-5-yl, indan-5-ylethan-1-one, 1-indane-5-yl-ethanone, acmc-1arc8, 1-2,3-dihydro-1h-inden-6-yl ethanone, 1-2,3-dihydro-1h-inden-5-yl-ethanone | |
| CC(=O)C1=CC2=C(CCC2)C=C1 | |
| 160.22 | |
| 160.22 | |
| Liquid |
| Yellow | |
| C11H12O | |
| 5 g | |
| HCMWPTFPUCPKQM-UHFFFAOYSA-N | |
| 1-(2,3-dihydro-1H-inden-5-yl)ethanone | |
| 233976 | |
| ≥98.0% (GC) | |
| 5-Acetylindan |
Safety and Handling
TSCA : No