accessibility menu, dialog, popup

UserName

CAS RN 4228-10-8

CH3O

CAS RN 4228-10-8

IUPAC Name: 1-(2,3-dihydro-1H-inden-5-yl)ethan-1-one
Synonyms: 1-(2,3-dihydro-1H-inden-5-yl)ethanone
Molecular Weight (g/mol): 160.22
Molecular Formula: C11H12O
InChi Key: HCMWPTFPUCPKQM-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C2CCCC2=C1
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)