missing translation for 'onlineSavingsMsg'
Learn More

5-Acetyl-2-chloropyridine 98.0+%, TCI America™

Catalog No. p-7118081
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27961G 1 g
A27965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27961G Supplier TCI America Supplier No. A27961G
Only null left

Chemical Identifiers

CAS 55676-22-7
Molecular Formula C7H6ClNO
Molecular Weight (g/mol) 155.58
MDL Number MFCD03407343
InChI Key UXSNZYGTQTXRAD-UHFFFAOYSA-N
Synonym 1-6-chloropyridin-3-yl ethanone, 2-chloro-5-acetylpyridine, 3-acetyl-6-chloropyridine, 5-acetyl-2-chloropyridine, 1-6-chloropyridin-3-yl ethan-1-one, 1-6-chloro-3-pyridinyl-1-ethanone, 6-chloro-3-acetylpyridine, 2-chloro-5-acetyl-pyridine, 1-6-chloro-3-pyridinyl ethanone, 1-6-chloro-pyridin-3-yl-ethanone
PubChem CID 2779698
IUPAC Name 1-(6-chloropyridin-3-yl)ethan-1-one
SMILES CC(=O)C1=CN=C(Cl)C=C1

Specifications

Melting Point 102°C
Quantity 1 g
Formula Weight 155.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Acetyl-2-chloropyridine