missing translation for 'onlineSavingsMsg'
Learn More

5-Acetyl-2,4-dimethylthiazole 98.0+%, TCI America™

Catalog No. p-7117312
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A121510G 10 g
A121525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A121510G Supplier TCI America Supplier No. A121510G
Only null left

Chemical Identifiers

CAS 38205-60-6
Molecular Formula C7H9NOS
Molecular Weight (g/mol) 155.215
MDL Number MFCD00005331
InChI Key BLQOKWQUTLNKON-UHFFFAOYSA-N
Synonym 5-acetyl-2,4-dimethylthiazole, 2,4-dimethyl-5-acetylthiazole, 1-2,4-dimethylthiazol-5-yl ethanone, 1-2,4-dimethyl-1,3-thiazol-5-yl ethanone, unii-pd36fu2wgy, ethanone, 1-2,4-dimethyl-5-thiazolyl, 1-2,4-dimethyl-5-thiazolyl ethanone, pd36fu2wgy, 2,4-dimethyl-5-thiazoyl methyl ketone, ketone, 2,4-dimethyl-5-thiazolyl methyl
PubChem CID 520888
IUPAC Name 1-(2,4-dimethyl-1,3-thiazol-5-yl)ethanone
SMILES CC1=C(SC(=N1)C)C(=O)C

Specifications

Boiling Point 230°C
Quantity 10 g
Formula Weight 155.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Acetyl-2,4-dimethylthiazole
EINECSNumber (5)-0209
TSCA No