accessibility menu, dialog, popup

UserName

CAS RN 38205-60-6

IUPAC Name: 1-(2,4-dimethyl-1,3-thiazol-5-yl)ethan-1-one
Synonyms: 1-(2,4-dimethyl-1,3-thiazol-5-yl)ethanone
Molecular Weight (g/mol): 155.22
Molecular Formula: C7H9NOS
InChi Key: BLQOKWQUTLNKON-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(C)N=C(C)S1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)