missing translation for 'onlineSavingsMsg'
Learn More

5-Acetyl-2,4-dimethylthiazole, 99%

Catalog No. p-7021419
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1374106 5 g
AAA1374114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1374106 Supplier Thermo Scientific Chemicals Supplier No. A1374106

CAS: 38205-60-6 | C7H9NOS | 155.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38205-60-6
Molecular Formula C7H9NOS
Molecular Weight (g/mol) 155.215
MDL Number MFCD00005331
InChI Key BLQOKWQUTLNKON-UHFFFAOYSA-N
Synonym 5-acetyl-2,4-dimethylthiazole, 2,4-dimethyl-5-acetylthiazole, 1-2,4-dimethylthiazol-5-yl ethanone, 1-2,4-dimethyl-1,3-thiazol-5-yl ethanone, unii-pd36fu2wgy, ethanone, 1-2,4-dimethyl-5-thiazolyl, 1-2,4-dimethyl-5-thiazolyl ethanone, pd36fu2wgy, 2,4-dimethyl-5-thiazoyl methyl ketone, ketone, 2,4-dimethyl-5-thiazolyl methyl
PubChem CID 520888
IUPAC Name 1-(2,4-dimethyl-1,3-thiazol-5-yl)ethanone
SMILES CC1=C(SC(=N1)C)C(=O)C

Specifications

Density 1.15
Boiling Point 230°C
Flash Point 104°C (219°F)
Refractive Index 1.543
Quantity 5 g
Beilstein 120560
Formula Weight 155.22
Percent Purity 99%
Chemical Name or Material 5-Acetyl-2,4-dimethylthiazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 253-826-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only