missing translation for 'onlineSavingsMsg'
Learn More

5-Acetamidopyridine-3-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog No. AAH66479MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66479MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH66479MD Supplier Thermo Scientific Chemicals Supplier No. H66479MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1201645-46-6
Molecular Formula C13H19BN2O3
Molecular Weight (g/mol) 262.116
MDL Number MFCD11878288
InChI Key ADPVUZOCOXRLMP-UHFFFAOYSA-N
Synonym n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-3-yl acetamide, 5-acetamido pyridine-3-boronic acid pinacol ester, 5-n-acetyl-amino-pyridin-3-ylboronic acid pinacol ester, 5-acetamidopyridin-3-yl boronic acid pinacol ester, n-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-3-yl acetamide, n-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-3-yl acetamide
PubChem CID 57415705
IUPAC Name N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl]acetamide
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CN=C2)NC(=O)C

Specifications

Quantity 250 mg
Beilstein 19927460
Formula Weight 262.12
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Acetamidopyridine-3-boronic acid pinacol ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only