accessibility menu, dialog, popup

UserName

CAS RN 1201645-46-6

IUPAC Name: N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl]acetamide
Synonyms: N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl]acetamide
Molecular Weight (g/mol): 262.12
Molecular Formula: C13H19BN2O3
InChi Key: ADPVUZOCOXRLMP-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CN=CC(=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results