missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5'-Acetamido-2'-hydroxyacetophenone 98.0+%, TCI America™
$82.79 - $281.08
Chemical Identifiers
| CAS | 7298-67-1 |
|---|---|
| Molecular Formula | C10H11NO3 |
| Molecular Weight (g/mol) | 193.202 |
| MDL Number | MFCD00100490 |
| InChI Key | DIQSYMRVTOVKQT-UHFFFAOYSA-N |
| Synonym | n-3-acetyl-4-hydroxyphenyl acetamide, n1-3-acetyl-4-hydroxyphenyl acetamide, n-3-acetyl-4-hydroxy-phenyl acetamide, 5'-acetamido-2'-hydroxyacetophenone, acetamide, n-3-acetyl-4-hydroxyphenyl, ksc380g7h, 3'-acetyl-4'-hydroxyacetanilide, 5-acetamido-2-hydroxyacetophenone, 5-acetamido-2-hydroxy-acetophenone, n-3-acetyl-4-hydroxyphenyl-acetamide |
| PubChem CID | 81720 |
| IUPAC Name | N-(3-acetyl-4-hydroxyphenyl)acetamide |
| SMILES | CC(=O)C1=C(C=CC(=C1)NC(=O)C)O |
Chemical Identifiers
| 7298-67-1 | |
| 193.202 | |
| DIQSYMRVTOVKQT-UHFFFAOYSA-N | |
| 81720 | |
| CC(=O)C1=C(C=CC(=C1)NC(=O)C)O |
| C10H11NO3 | |
| MFCD00100490 | |
| n-3-acetyl-4-hydroxyphenyl acetamide, n1-3-acetyl-4-hydroxyphenyl acetamide, n-3-acetyl-4-hydroxy-phenyl acetamide, 5'-acetamido-2'-hydroxyacetophenone, acetamide, n-3-acetyl-4-hydroxyphenyl, ksc380g7h, 3'-acetyl-4'-hydroxyacetanilide, 5-acetamido-2-hydroxyacetophenone, 5-acetamido-2-hydroxy-acetophenone, n-3-acetyl-4-hydroxyphenyl-acetamide | |
| N-(3-acetyl-4-hydroxyphenyl)acetamide |
Specifications
| 7298-67-1 | |
| C10H11NO3 | |
| 1 g | |
| DIQSYMRVTOVKQT-UHFFFAOYSA-N | |
| N-(3-acetyl-4-hydroxyphenyl)acetamide | |
| 81720 | |
| ≥98.0% (HPLC,T) | |
| 5′-Acetamido-2′-hydroxyacetophenone |
| 168°C | |
| MFCD00100490 | |
| n-3-acetyl-4-hydroxyphenyl acetamide, n1-3-acetyl-4-hydroxyphenyl acetamide, n-3-acetyl-4-hydroxy-phenyl acetamide, 5'-acetamido-2'-hydroxyacetophenone, acetamide, n-3-acetyl-4-hydroxyphenyl, ksc380g7h, 3'-acetyl-4'-hydroxyacetanilide, 5-acetamido-2-hydroxyacetophenone, 5-acetamido-2-hydroxy-acetophenone, n-3-acetyl-4-hydroxyphenyl-acetamide | |
| CC(=O)C1=C(C=CC(=C1)NC(=O)C)O | |
| 193.202 | |
| 193.20 | |
| Crystalline Powder |
Safety and Handling
TSCA : No