missing translation for 'onlineSavingsMsg'
Learn More

5-Acetamido-2-aminobenzoic acid, 97%

Catalog No. AAL1125606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1125606 5 g
AAL1125614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1125606 Supplier Thermo Scientific Chemicals Supplier No. L1125606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50670-83-2 | C9H10N2O3 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50670-83-2
Molecular Formula C9H10N2O3
Molecular Weight (g/mol) 194.19
MDL Number MFCD00060120
InChI Key GSOHXJQXAKNJES-UHFFFAOYSA-N
Synonym 5-acetamidoanthranilic acid, 2-amino-5-acetamidobenzoic acid, benzoic acid, 5-acetylamino-2-amino, 5-acetylamino-2-aminobenzoic acid, 5-acetamino-2-aminobenzoic acid, 5-acetamidoanthranilicacid, acmc-209kp2, 5-acetylaminoanthanilic acid, 5-acetamido-2-aminobenzoicacid, 2-amino-5-acetaminobenzoic acid
PubChem CID 170890
IUPAC Name 5-acetamido-2-aminobenzoic acid
SMILES CC(=O)NC1=CC(=C(C=C1)N)C(=O)O

Specifications

Melting Point ∽230°C (decomposition)
Quantity 5 g
Beilstein 2726882
Formula Weight 194.19
Percent Purity 97%
Chemical Name or Material 5-Acetamido-2-aminobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 256-702-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only