missing translation for 'onlineSavingsMsg'
Learn More

5,8-Dibromo-1,2,4-triazolo[1,5-a]pyrazine, 95%, Thermo Scientific Chemicals

Catalog No. p-7040067
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5968103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5968103 Supplier Thermo Scientific Chemicals Supplier No. H5968103
Only null left

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 959755-46-5
Molecular Formula C5H2Br2N4
Molecular Weight (g/mol) 277.907
MDL Number MFCD11040378
InChI Key IDWOTLNDEITXRA-UHFFFAOYSA-N
Synonym 5,8-dibromo-1,2,4 triazolo 1,5-a pyrazine, 5,8-dibromo 1,2,4 triazolo 1,5-a pyrazine, 5,8-dibromo-1,2,4-triazolo 1,5-a pyrazine, 5,8-dibromo 1,2,4 triazolo 1,5-a-pyrazine
PubChem CID 49758022
IUPAC Name 5,8-dibromo-[1,2,4]triazolo[1,5-a]pyrazine
SMILES C1=C(N2C(=NC=N2)C(=N1)Br)Br

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 277.91
Percent Purity 95%
Chemical Name or Material 5,8-Dibromo-1,2,4-triazolo[1,5-a]pyrazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only