CAS RN 959755-46-5
IUPAC Name:
5,8-dibromo-[1,2,4]triazolo[1,5-a]pyrazine
Synonyms:
5,8-dibromo-[1,2,4]triazolo[1,5-a]pyrazine
Molecular Weight (g/mol):
277.91
Molecular Formula:
C5H2Br2N4
InChi Key:
IDWOTLNDEITXRA-UHFFFAOYSA-N
SMILES:
BrC1=CN=C(Br)C2=NC=NN12
1
–
1
of
1
results
1
Promotions Available
5,8-Dibromo-1,2,4-triazolo[1,5-a]pyrazine, 95%, Thermo Scientific Chemicals
5,8-Dibromo-1,2,4-triazolo[1,5-a]pyrazine, 95%, Thermo Scientific Chemicals
CAS: 959755-46-5 Molecular Formula: C5H2Br2N4 Molecular Weight (g/mol): 277.907 MDL Number: MFCD11040378 InChI Key: IDWOTLNDEITXRA-UHFFFAOYSA-N Synonym: 5,8-dibromo-1,2,4 triazolo 1,5-a pyrazine,5,8-dibromo 1,2,4 triazolo 1,5-a pyrazine,5,8-dibromo-1,2,4-triazolo 1,5-a pyrazine,5,8-dibromo 1,2,4 triazolo 1,5-a-pyrazine PubChem CID: 49758022 IUPAC Name: 5,8-dibromo-[1,2,4]triazolo[1,5-a]pyrazine SMILES: C1=C(N2C(=NC=N2)C(=N1)Br)Br