missing translation for 'onlineSavingsMsg'
Learn More

5,7-Difluoro-1-indanone, 97%

Catalog No. p-7035794
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5400203 1 g
AAH5400206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5400203 Supplier Thermo Scientific Chemicals Supplier No. H5400203

CAS: 84315-25-3 | C9H6F2O | 168.14 g/mol

5,7-Difluoro-1-indanone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,7-Difluoro-1-indanone is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water 0.081 g/L 25°C.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 84315-25-3
Molecular Formula C9H6F2O
Molecular Weight (g/mol) 168.14
MDL Number MFCD09908161
InChI Key XWZGNWCKXLECBO-UHFFFAOYSA-N
Synonym 5,7-difluoro-1-indanone, 5,7-difluoro-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 5,7-difluoro-2,3-dihydro, 5,7-difluoroindan-1-one, 1h-inden-1-one,5,7-difluoro-2,3-dihydro, acmc-20a83k, 5,7-difluoro-2,3-dihydro-1h-indene-1-one
PubChem CID 11789595
IUPAC Name 5,7-difluoro-2,3-dihydroinden-1-one
SMILES FC1=CC(F)=C2C(=O)CCC2=C1

Specifications

Melting Point 81°C
Quantity 1 g
Solubility Information Sparingly soluble in water 0.081g/L 25°C.
Formula Weight 168.14
Percent Purity 97%
Chemical Name or Material 5,7-Difluoro-1-indanone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only