missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dichloro-1-indanone, 97%

Catalog No. p-7035799
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5400703 1 g
AAH5400706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5400703 Supplier Thermo Scientific Chemicals Supplier No. H5400703
Only null left

CAS: 448193-94-0 | C9H6Cl2O | 201.05 g/mol

5,7-Dichloro-1-indanone is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,7-Dichloro-1-indanone is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 448193-94-0
Molecular Formula C9H6Cl2O
Molecular Weight (g/mol) 201.05
MDL Number MFCD09908160
InChI Key TWNXYJNWNZZECV-UHFFFAOYSA-N
Synonym 5,7-dichloro-2,3-dihydro-1h-inden-1-one, 5,7-dichloro-1-indanone, 1h-inden-1-one, 5,7-dichloro-2,3-dihydro, 5,7-dichloroindan-1-one, acmc-1asf2
PubChem CID 22292346
SMILES ClC1=CC(Cl)=C2C(=O)CCC2=C1

Specifications

Melting Point 98°C
Quantity 1 g
Solubility Information Slightly soluble in water.
IUPAC Name 5,7-dichloro-2,3-dihydro-1H-inden-1-one
Formula Weight 201.05
Percent Purity 97%
Chemical Name or Material 5,7-Dichloro-1-indanone
Hazard Category H302-H319
Hazard Statement GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only