accessibility menu, dialog, popup

UserName

CAS RN 448193-94-0

IUPAC Name: 5,7-dichloro-2,3-dihydro-1H-inden-1-one
Synonyms: 5,7-dichloro-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 201.05
Molecular Formula: C9H6Cl2O
InChi Key: TWNXYJNWNZZECV-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C2C(=O)CCC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results