missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dichloro-1,6-naphthyridine, 97%

Catalog No. p-4229168
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC440670010 1 g Glass bottle
AC440670050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC440670010 Supplier Thermo Scientific Chemicals Supplier No. 440670010

CAS: 337958-60-8 | C8H4Cl2N2 | 199.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 337958-60-8
Molecular Formula C8H4Cl2N2
Molecular Weight (g/mol) 199.04
InChI Key HNRPBMNTCYRAJD-UHFFFAOYSA-N
Synonym pubchem14653, 5,7-dichloro-1,6-napthyridine, 1,6-naphthyridine,5,7-dichloro, 1,6-naphthyridine, 5,7-dichloro, 5,7-bis chloranyl-1,6-naphthyridine
PubChem CID 12204233
IUPAC Name 5,7-dichloro-1,6-naphthyridine
SMILES C1=CC2=C(N=C(C=C2N=C1)Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 138°C to 140°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 199.04
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5, 7-Dichloro-1, 6-naphthyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

RUO – Research Use Only