accessibility menu, dialog, popup

UserName

CAS RN 337958-60-8

IUPAC Name: 5,7-dichloro-1,6-naphthyridine
Synonyms: 5,7-dichloro-1,6-naphthyridine
Molecular Weight (g/mol): 199.03
Molecular Formula: C8H4Cl2N2
InChi Key: HNRPBMNTCYRAJD-UHFFFAOYSA-N
SMILES: ClC1=CC2=NC=CC=C2C(Cl)=N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results