missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dibromo-1-indanone, 97%

Catalog No. p-7035797
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5400503 1 g
AAH5400506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5400503 Supplier Thermo Scientific Chemicals Supplier No. H5400503
Only null left

CAS: 923977-18-8 | C9H6Br2O | 289.954 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 923977-18-8
Molecular Formula C9H6Br2O
Molecular Weight (g/mol) 289.954
MDL Number MFCD11707100
InChI Key WLRPGDOWFWBUAE-UHFFFAOYSA-N
Synonym 5,7-dibromo-1-indanone, 5,7-dibromo-2,3-dihydro-1h-inden-1-one, 5,7-dibromoindanone, 5,7-dibromoindan-1-one, 5,7-bis bromanyl-2,3-dihydroinden-1-one
PubChem CID 16038257
IUPAC Name 5,7-dibromo-2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=C(C=C(C=C21)Br)Br

Specifications

Melting Point 148°C
Quantity 1 g
Formula Weight 289.95
Percent Purity 97%
Chemical Name or Material 5,7-Dibromo-1-indanone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only