missing translation for 'onlineSavingsMsg'
Learn More

5,7,12,14-Pentacenetetrone 95.0+%, TCI America™

Catalog No. p-7135219
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17105G 5 g
P171025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17105G Supplier TCI America Supplier No. P17105G
Only null left

Chemical Identifiers

CAS 23912-79-0
Molecular Formula C22H10O4
Molecular Weight (g/mol) 338.318
MDL Number MFCD00075209
InChI Key YZOGOBWHTVNKGA-UHFFFAOYSA-N
Synonym 5,7,12,14-pentacenetetrone, 2.3-phthalylanthrachinon, pentacene-5,7,12,14-tetraone, 5,7,12,14-tetraoxo-5,7,12,14-tetrahydropentacene, 5,7,12,14-tetrahydropentacene-5,7,12,14-tetrone
PubChem CID 4733
IUPAC Name pentacene-5,7,12,14-tetrone
SMILES C1=CC=C2C(=C1)C(=O)C3=CC4=C(C=C3C2=O)C(=O)C5=CC=CC=C5C4=O

Specifications

Melting Point 400°C
Color White-Yellow
Quantity 5 g
Formula Weight 338.32
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Lumps
Chemical Name or Material 5,7,12,14-Pentacenetetrone