missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 5(6)-TAMRA-X, SE (6-(Tetramethylrhodamine-5-(and-6)-Carboxamido) Hexanoic Acid, Succinimidyl Ester), mixed isomers

Catalog No. T6105
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T6105 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T6105 Supplier Invitrogen™ Supplier No. T6105

5(6)-TAMRA-X, SE (6-(Tetramethylrhodamine-5-(and-6)-Carboxamido) Hexanoic Acid, Succinimidyl Ester), mixed isomers

The amine-reactive 6-(tetramethylrhodamine-5-(and-6)-carboxamido)hexanoic acid, succinimidyl ester (5(6)-TAMRA-X, SE) can be used to can be used to create bright red-orange fluorescent bioconjugates with excitation/emission maxima ∼555/580 nm.

Specifications

Quantity 10 mg
Product Type 5(6)-TAMRA-X SE
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye TAMRA™ Isomers, TMR (Tetramethylrhodamine)
Label Type Classic Dyes
Emission 580
Excitation 555

For Research Use Only. Not for use in diagnostic procedures.