missing translation for 'onlineSavingsMsg'
Learn More

(+)-5,6-O-Isopropylidene-L-ascorbic Acid 98.0+%, TCI America™

Catalog No. p-7131450
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05075G 5 g
I050725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05075G Supplier TCI America Supplier No. I05075G
Only null left

Chemical Identifiers

CAS 15042-01-0
Molecular Formula C9H12O6
Molecular Weight (g/mol) 216.189
MDL Number MFCD00010552
InChI Key POXUQBFHDHCZAD-MHTLYPKNSA-N
PubChem CID 54691418
IUPAC Name (2R)-2-[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]-3,4-dihydroxy-2H-furan-5-one
SMILES CC1(OCC(O1)C2C(=C(C(=O)O2)O)O)C

Specifications

Melting Point 210°C
Color White-Yellow
Quantity 5 g
Formula Weight 216.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (+)-5,6-O-Isopropylidene-L-ascorbic Acid