missing translation for 'onlineSavingsMsg'
Learn More

5,6-Methylenedioxy-2-phenylindole, 97%

Catalog No. p-7025535
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2106503 1 g
AAB2106506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2106503 Supplier Thermo Scientific Chemicals Supplier No. B2106503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 64943-90-4 | C15H11NO2 | 237.258 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64943-90-4
Molecular Formula C15H11NO2
Molecular Weight (g/mol) 237.258
MDL Number MFCD00798602
InChI Key OURPDRQDIRKULF-UHFFFAOYSA-N
Synonym 5,6-methylenedioxy-2-phenylindole, 6-phenyl-5h-1,3 dioxolo 4,5-f indole, 6-phenyl-2h,5h-1,3 dioxolo 4,5-f indole, acmc-20am77, 6-phenyl-5h-1,3-dioxolo 4,5-f indole, 5h-1,3-dioxolo 4,5-f indole,6-phenyl, 6-phenyl-2h-1,3-dioxolano 4,5-f indole
PubChem CID 4067102
IUPAC Name 6-phenyl-5H-[1,3]dioxolo[4,5-f]indole
SMILES C1OC2=C(O1)C=C3C(=C2)C=C(N3)C4=CC=CC=C4

Specifications

Quantity 1 g
Formula Weight 237.26
Percent Purity 97%
Chemical Name or Material 5,6-Methylenedioxy-2-phenylindole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only