CAS RN 64943-90-4
IUPAC Name:
6-phenyl-2H,5H-[1,3]dioxolo[4,5-f]indole
Synonyms:
6-phenyl-2H,5H-[1,3]dioxolo[4,5-f]indole
Molecular Weight (g/mol):
237.26
Molecular Formula:
C15H11NO2
InChi Key:
OURPDRQDIRKULF-UHFFFAOYSA-N
SMILES:
C1OC2=C(O1)C=C1C=C(NC1=C2)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
5,6-Methylenedioxy-2-phenylindole, 97%
5,6-Methylenedioxy-2-phenylindole, 97%
CAS: 64943-90-4 Molecular Formula: C15H11NO2 Molecular Weight (g/mol): 237.258 MDL Number: MFCD00798602 InChI Key: OURPDRQDIRKULF-UHFFFAOYSA-N Synonym: 5,6-methylenedioxy-2-phenylindole,6-phenyl-5h-1,3 dioxolo 4,5-f indole,6-phenyl-2h,5h-1,3 dioxolo 4,5-f indole,acmc-20am77,6-phenyl-5h-1,3-dioxolo 4,5-f indole,5h-1,3-dioxolo 4,5-f indole,6-phenyl,6-phenyl-2h-1,3-dioxolano 4,5-f indole PubChem CID: 4067102 IUPAC Name: 6-phenyl-5H-[1,3]dioxolo[4,5-f]indole SMILES: C1OC2=C(O1)C=C3C(=C2)C=C(N3)C4=CC=CC=C4