missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dihydrouracil, 97%

Catalog No. AAL0191803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0191803 1 g
AAL0191806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0191803 Supplier Thermo Scientific Chemicals Supplier No. L0191803

CAS: 504-07-4 | C4H6N2O2 | 114.10 g/mol

5,6-Dihydrouracil acts as an intermediate in the catabolism of uracil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,6-Dihydrouracil acts as an intermediate in the catabolism of uracil.

Solubility
Soluble in Sodium Hydroxide.

Notes
Incompatible with oxidizing agents.Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 504-07-4
Molecular Formula C4H6N2O2
Molecular Weight (g/mol) 114.10
MDL Number MFCD00006029
InChI Key OIVLITBTBDPEFK-UHFFFAOYSA-N
Synonym dihydrouracil, 5,6-dihydrouracil, hydrouracil, 5,6-dihydro-2,4-dihydroxypyrimidine, dihydropyrimidine-2,4 1h,3h-dione, dihydrouracile, 2,4 1h,3h-pyrimidinedione, dihydro, di-h-uracil, dihydro-2,4 1h,3h-pyrimidinedione, hydrouracil 8ci
PubChem CID 649
ChEBI CHEBI:15901
IUPAC Name 1,3-diazinane-2,4-dione
SMILES O=C1CCNC(=O)N1

Specifications

Melting Point 278°C to 280°C
Quantity 1 g
Beilstein 112496
Solubility Information Soluble in Sodium Hydroxide.
Formula Weight 114.1
Percent Purity 97%
Chemical Name or Material 5,6-Dihydrouracil
EINECSNumber 207-982-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only