CAS RN 504-07-4
IUPAC Name:
1,3-diazinane-2,4-dione
Synonyms:
dihydrouracil|3,4-dihydrouracil
Molecular Weight (g/mol):
114.1
Molecular Formula:
C4H6N2O2
InChi Key:
OIVLITBTBDPEFK-UHFFFAOYSA-N
SMILES:
O=C1CCNC(=O)N1
1
–
1
of
1
results
1
5,6-Dihydrouracil, 97%
CAS: 504-07-4 Molecular Formula: C4H6N2O2 Molecular Weight (g/mol): 114.10 MDL Number: MFCD00006029 InChI Key: OIVLITBTBDPEFK-UHFFFAOYSA-N Synonym: dihydrouracil,5,6-dihydrouracil,hydrouracil,5,6-dihydro-2,4-dihydroxypyrimidine,dihydropyrimidine-2,4 1h,3h-dione,dihydrouracile,2,4 1h,3h-pyrimidinedione, dihydro,di-h-uracil,dihydro-2,4 1h,3h-pyrimidinedione,hydrouracil 8ci PubChem CID: 649 ChEBI: CHEBI:15901 IUPAC Name: 1,3-diazinane-2,4-dione SMILES: O=C1CCNC(=O)N1