missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dihydro-1,4-dithiin-2,3-dicarboximide 97.0+%, TCI America™

Catalog No. D25745G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25745G 5 g
D25741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25745G Supplier TCI America Supplier No. D25745G
Only null left

Chemical Identifiers

CAS 24519-85-5
Molecular Formula C6H5NO2S2
Molecular Weight (g/mol) 187.23
MDL Number MFCD01632185
InChI Key XNIVQWHLYNEVBN-UHFFFAOYSA-N
Synonym 2,3-Dihydro-1,4-dithiino[2,3-c]pyrrole-5,7-dione
PubChem CID 32477
IUPAC Name 2H,3H,5H,6H,7H-[1,4]dithiino[2,3-c]pyrrole-5,7-dione
SMILES O=C1NC(=O)C2=C1SCCS2

Specifications

Melting Point 217°C
Quantity 5 g
Formula Weight 187.23
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dihydro-1,4-dithiin-2,3-dicarboximide
RTECSNumber JO6955000
TSCA No