accessibility menu, dialog, popup

UserName

CAS RN 24519-85-5

OHNOSS

CAS RN 24519-85-5

IUPAC Name: 2H,3H,5H,6H,7H-[1,4]dithiino[2,3-c]pyrrole-5,7-dione
Synonyms: 2H,3H,6H-[1,4]dithiino[2,3-c]pyrrole-5,7-dione
Molecular Weight (g/mol): 187.23
Molecular Formula: C6H5NO2S2
InChi Key: XNIVQWHLYNEVBN-UHFFFAOYSA-N
SMILES: O=C1NC(=O)C2=C1SCCS2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results