missing translation for 'onlineSavingsMsg'
Learn More

5,6-Difluoro-o-anisaldehyde 97.0+%, TCI America™

Catalog No. D38855G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38855G 5 g
D38851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38855G Supplier TCI America Supplier No. D38855G

Chemical Identifiers

CAS 187543-87-9
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD02093968
InChI Key AKOJAYHBKACKNJ-UHFFFAOYSA-N
Synonym 5,6-difluoro-o-anisaldehyde, 2,3-difluoro-6-methoxy-benzaldehyde, 3,4-difluoro-2-formylanisole, pubchem2902, acmc-1bwkg, 2-methoxy-5,6-difluorobenzaldehyde, 2-methoxy-5,6-difluoro-benzaldehyde, 2,3-difluoro-6-methyloxy benzaldehyde, benzaldehyde, 2,3-difluoro-6-methoxy, 2,3-bis fluoranyl-6-methoxy-benzaldehyde
PubChem CID 2774112
IUPAC Name 2,3-difluoro-6-methoxybenzaldehyde
SMILES COC1=C(C(=C(C=C1)F)F)C=O

Specifications

Melting Point 58°C
Quantity 5 g
Formula Weight 172.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Difluoro-o-anisaldehyde