missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dichloroindole 98.0+%, TCI America™

Catalog No. D49245G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49245G 5 g
D49241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49245G Supplier TCI America Supplier No. D49245G
Only null left

Chemical Identifiers

CAS 121859-57-2
Molecular Formula C8H5Cl2N
Molecular Weight (g/mol) 186.04
MDL Number MFCD01075036
InChI Key ILINOHVVKWYAFM-UHFFFAOYSA-N
PubChem CID 10487776
IUPAC Name 5,6-dichloro-1H-indole
SMILES ClC1=C(Cl)C=C2C=CNC2=C1

Specifications

Melting Point 152°C
Quantity 5 g
Formula Weight 186.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dichloroindole