missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dichloro-2-methylbenzimidazole 98.0+%, TCI America™

Catalog No. d34881g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
D34881G 1 g
D34885G 5 g
2 options

Catalog No. D34881G

Supplier: TCI America D34881G

Only null left

Chemical Identifiers

CAS 6478-79-1
Molecular Formula C8H6Cl2N2
Molecular Weight (g/mol) 201.05
MDL Number MFCD00276973
InChI Key WMOBNOCVMZFPEN-UHFFFAOYSA-N
PubChem CID 608319
IUPAC Name 5,6-dichloro-2-methyl-1H-1,3-benzodiazole
SMILES CC1=NC2=CC(Cl)=C(Cl)C=C2N1

Specifications

Melting Point 248°C
Color White-Yellow
Quantity 1 g
Formula Weight 201.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dichloro-2-methylbenzimidazole