missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dichloro-2,3-dicyanopyrazine 98.0+%, TCI America™

Catalog No. D227110G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D227110G 10 g
D227125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D227110G Supplier TCI America Supplier No. D227110G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56413-95-7
Molecular Formula C6Cl2N4
Molecular Weight (g/mol) 198.99
MDL Number MFCD00191419
InChI Key QUFXYBKGILUJHS-UHFFFAOYSA-N
Synonym 5,6-Dichloro-2,3-pyrazinedicarbonitrile
PubChem CID 607820
IUPAC Name dichloropyrazine-2,3-dicarbonitrile
SMILES ClC1=NC(C#N)=C(N=C1Cl)C#N

Specifications

Melting Point 183°C
Quantity 10 g
UN Number 2811
Formula Weight 198.99
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dichloro-2,3-dicyanopyrazine