accessibility menu, dialog, popup

UserName

CAS RN 56413-95-7

IUPAC Name: dichloropyrazine-2,3-dicarbonitrile
Synonyms: dichloropyrazine-2,3-dicarbonitrile
Molecular Weight (g/mol): 198.99
Molecular Formula: C6Cl2N4
InChi Key: QUFXYBKGILUJHS-UHFFFAOYSA-N
SMILES: ClC1=NC(C#N)=C(N=C1Cl)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results