missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dichloro-1-ethyl-2-methylbenzimidazole 98.0+%, TCI America™

Catalog No. D25565G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D25565G Supplier TCI America Supplier No. D25565G

Chemical Identifiers

CAS 3237-62-5
Molecular Formula C10H10Cl2N2
Molecular Weight (g/mol) 229.10
MDL Number MFCD00022843
InChI Key IVVLMQPTQOLYDX-UHFFFAOYSA-N
Synonym 5,6-dichloro-1-ethyl-2-methyl-1h-benzo d imidazole, 1-ethyl-2-methyl-5,6-dichlorobenzimidazole, 5,6-dichloro-1-ethyl-2-methyl-1h-benzimidazole, 1h-benzimidazole, 5,6-dichloro-1-ethyl-2-methyl, dichloroethylmethylbenzimidazole, 5,6-dichloro-1-ethyl-2-methyl-1,3-benzodiazole, 5,6-dichloro-1-ethyl-2-methyl-1h-benzoimidazole, 5,6-dichloro-1-ethyl-2-methyl-1h-1,3-benzodiazole, pubchem7601, acmc-1cr6w
PubChem CID 76717
IUPAC Name 5,6-dichloro-1-ethyl-2-methyl-1H-1,3-benzodiazole
SMILES CCN1C(C)=NC2=CC(Cl)=C(Cl)C=C12

Specifications

Melting Point 110°C
Boiling Point 195°C
Quantity 5 g
Formula Weight 229.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dichloro-1-ethyl-2-methylbenzimidazole
EINECSNumber (5)-0464
TSCA Yes