accessibility menu, dialog, popup

UserName

H3CNH3CNClCl

CAS RN 3237-62-5

IUPAC Name: 5,6-dichloro-1-ethyl-2-methyl-1H-1,3-benzodiazole
Synonyms: 5,6-dichloro-1-ethyl-2-methyl-1,3-benzodiazole
Molecular Weight (g/mol): 229.1
Molecular Formula: C10H10Cl2N2
InChi Key: IVVLMQPTQOLYDX-UHFFFAOYSA-N
SMILES: CCN1C(C)=NC2=CC(Cl)=C(Cl)C=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results