missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dibromo-2,1,3-benzothiadiazole 98.0+%, TCI America™

Catalog No. D40645G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D40645G Supplier TCI America Supplier No. D40645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18392-81-9
Molecular Formula C6H2Br2N2S
Molecular Weight (g/mol) 293.96
MDL Number MFCD20257910
InChI Key XIDGOHYQUCNHTL-UHFFFAOYSA-N
PubChem CID 11044589
IUPAC Name 5,6-dibromo-2,1,3-benzothiadiazole
SMILES BrC1=CC2=NSN=C2C=C1Br

Specifications

Melting Point 133°C
Color Yellow
Quantity 5 g
Formula Weight 293.96
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dibromo-2,1,3-benzothiadiazole