accessibility menu, dialog, popup

UserName

NSNBrBr

CAS RN 18392-81-9

IUPAC Name: 5,6-dibromo-2,1,3-benzothiadiazole
Synonyms: 5,6-dibromo-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 293.96
Molecular Formula: C6H2Br2N2S
InChi Key: XIDGOHYQUCNHTL-UHFFFAOYSA-N
SMILES: BrC1=CC2=NSN=C2C=C1Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results