missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,6-Dibromo-1-hexene 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 55°C |
| Quantity | 5 g |
| Formula Weight | 241.95 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 5,6-Dibromo-1-hexene |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |