missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dibromo-1-hexene 96.0+%, TCI America™

Catalog No. D48441G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48441G 1 g
D48445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48441G Supplier TCI America Supplier No. D48441G
Only null left

Chemical Identifiers

CAS 4285-48-7
Molecular Formula C6H10Br2
Molecular Weight (g/mol) 241.954
MDL Number MFCD00041698
InChI Key WFGDPUOTONXJOS-UHFFFAOYSA-N
PubChem CID 2755398
IUPAC Name 5,6-dibromohex-1-ene
SMILES C=CCCC(CBr)Br

Specifications

Color Yellow
Boiling Point 55°C
Quantity 1 g
Formula Weight 241.95
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 5,6-Dibromo-1-hexene
TSCA No
Recommended Storage Refrigerator