missing translation for 'onlineSavingsMsg'
Learn More

5,6-Diamino-1,3-dipropyluracil Hydrochloride 98.0+%, TCI America™

Catalog No. D48101G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D48101G Supplier TCI America Supplier No. D48101G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 324002-49-5
Molecular Formula C10H19ClN4O2
Molecular Weight (g/mol) 262.738
MDL Number MFCD06797515
InChI Key DWPGKJBUHDOVAD-UHFFFAOYSA-N
PubChem CID 18767618
IUPAC Name 5,6-diamino-1,3-dipropylpyrimidine-2,4-dione;hydrochloride
SMILES CCCN1C(=C(C(=O)N(C1=O)CCC)N)N.Cl

Specifications

Melting Point 244°C
Quantity 1 g
Formula Weight 262.74
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Diamino-1,3-dipropyluracil Hydrochloride