accessibility menu, dialog, popup

UserName

CH3NONOH3CNH2NH2HCl

CAS RN 324002-49-5

IUPAC Name: 5,6-diamino-1,3-dipropyl-1,2,3,4-tetrahydropyrimidine-2,4-dione hydrochloride
Synonyms: 5,6-diamino-1,3-dipropylpyrimidine-2,4-dione hydrochloride
Molecular Weight (g/mol): 262.74
Molecular Formula: C10H19ClN4O2
InChi Key: DWPGKJBUHDOVAD-UHFFFAOYSA-N
SMILES: Cl.CCCN1C(N)=C(N)C(=O)N(CCC)C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results