missing translation for 'onlineSavingsMsg'
Learn More

5,6,7-Trihydroxyflavone, 97%

Catalog No. AAL14185MC
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14185MC 100 mg
AAL14185ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL14185MC Supplier Thermo Scientific Chemicals Supplier No. L14185MC
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 491-67-8 | C15H10O5 | 270.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 491-67-8
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.24
MDL Number MFCD00017459
InChI Key FXNFHKRTJBSTCS-UHFFFAOYSA-N
Synonym baicalein, 5,6,7-trihydroxyflavone, noroxylin, 5,6,7-trihydroxy-2-phenyl-4h-chromen-4-one, biacalein, baicelein, chembl8260, 4h-1-benzopyran-4-one, 5,6,7-trihydroxy-2-phenyl, 5,7-trihydroxyflavone, 5,6,7-trihydroxy-2-phenyl-chromen-4-one
PubChem CID 5281605
ChEBI CHEBI:2979
IUPAC Name 5,6,7-trihydroxy-2-phenylchromen-4-one
SMILES OC1=C(O)C(O)=C2C(=O)C=C(OC2=C1)C1=CC=CC=C1

Specifications

Melting Point 268°C to 270°C
Quantity 100 mg
Beilstein 272683
Merck Index 14,942
Formula Weight 270.24
Percent Purity 97%
Chemical Name or Material 5,6,7-Trihydroxyflavone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only