missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetrahydroquinoline 96.0+%, TCI America™

Catalog No. t1150100ml
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1150100ML 100 mL
T115010ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1150100ML Supplier TCI America Supplier No. T1150100ML
Only null left

Chemical Identifiers

CAS 10500-57-9
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.194
MDL Number MFCD00006734
InChI Key YQDGQEKUTLYWJU-UHFFFAOYSA-N
Synonym 2,3-cyclohexeno pyridine, quinoline, 5,6,7,8-tetrahydro, unii-l786aag56h, 5,6,7,8-tetra-hydroquinoline, 5,6,7,8-tetrahydro-quinoline, pubchem5891, 2,3-cyclohexenopyridine, g00034-watson-int, 2,3-cyclohexano pyridine, acmc-2098eh
PubChem CID 66335
IUPAC Name 5,6,7,8-tetrahydroquinoline
SMILES C1CCC2=C(C1)C=CC=N2

Specifications

Color Yellow
Boiling Point 222°C
Quantity 100 mL
Formula Weight 133.19
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5,6,7,8-Tetrahydroquinoline
EINECSNumber (5)-6259
TSCA No