accessibility menu, dialog, popup

UserName

N

CAS RN 10500-57-9

IUPAC Name: 5,6,7,8-tetrahydroquinoline
Synonyms: quinoline, 5,6,7,8-tetrahydro-
Molecular Weight (g/mol): 133.19
Molecular Formula: C9H11N
InChi Key: YQDGQEKUTLYWJU-UHFFFAOYSA-N
SMILES: C1CCC2=NC=CC=C2C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)