missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetrahydroquinoline, 98%

Catalog No. AC188870250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC188870250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC188870250 Supplier Thermo Scientific Chemicals Supplier No. 188870250
Only null left

CAS: 10500-57-9 | C9H11N | 133.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10500-57-9
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
InChI Key YQDGQEKUTLYWJU-UHFFFAOYSA-N
Synonym 2,3-cyclohexeno pyridine, quinoline, 5,6,7,8-tetrahydro, unii-l786aag56h, 5,6,7,8-tetra-hydroquinoline, 5,6,7,8-tetrahydro-quinoline, pubchem5891, 2,3-cyclohexenopyridine, g00034-watson-int, 2,3-cyclohexano pyridine, acmc-2098eh
PubChem CID 66335
IUPAC Name 5,6,7,8-tetrahydroquinoline
SMILES C1CCC2=C(C1)C=CC=N2

Specifications

Density 1.0300g/mL
Boiling Point 218.0°C to 222.0°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Refractive Index 1.544
Quantity 25 g
Beilstein 20,II,176
Specific Gravity 1.03
Formula Weight 133.19
Percent Purity 98%
Chemical Name or Material 5,6,7,8-Tetrahydroquinoline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 234-030-2

RUO – Research Use Only