missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetrahydronaphthalene-1-carboxylic Acid 98.0+%, TCI America™

Catalog No. T267225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T267225G 25 g
T26725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T267225G Supplier TCI America Supplier No. T267225G

Chemical Identifiers

CAS 4242-18-6
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.22
MDL Number MFCD01464030
InChI Key GCFQXKYHWFWGSB-UHFFFAOYSA-N
PubChem CID 239604
IUPAC Name 5,6,7,8-tetrahydronaphthalene-1-carboxylic acid
SMILES OC(=O)C1=CC=CC2=C1CCCC2

Specifications

Melting Point 152°C
Color White
Quantity 25 g
Formula Weight 176.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5,6,7,8-Tetrahydronaphthalene-1-carboxylic Acid