missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetrahydro-2-naphthol 98.0+%, TCI America™

Catalog No. T010825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T010825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T010825G Supplier TCI America Supplier No. T010825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 1125-78-6
Melting Point 62°C
Boiling Point 133°C
Molecular Formula C10H12O
Quantity 25 g
MDL Number MFCD00001738
Synonym 5,6,7,8-tetrahydro-2-naphthol, 6-tetralinol, 6-hydroxytetralin, 2-naphthalenol, 5,6,7,8-tetrahydro, 5,6,7,8-tetrahydro-beta-naphthol, ac-beta-tetralol, 5,6,7,8-tetrahydro-2-hydroxynaphthalene, 2-naphthol, 5,6,7,8-tetrahydro, unii-tmr02i7n8s, 5,6,7,8-tetrahydro-2-naphthalenol
InChI Key UMKXSOXZAXIOPJ-UHFFFAOYSA-N
SMILES C1CCC2=C(C1)C=CC(=C2)O
IUPAC Name 5,6,7,8-tetrahydronaphthalen-2-ol
Molecular Weight (g/mol) 148.205
PubChem CID 14305
ChEBI CHEBI:34448
Formula Weight 148.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,6,7,8-Tetrahydro-2-naphthol
Show More Show Less
RTECSNumber QL5300000
TSCA No