missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetrahydro-2-naphthol, 98%

Catalog No. p-7045294
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1016406 5 g
AAL1016414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1016406 Supplier Thermo Scientific Chemicals Supplier No. L1016406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1125-78-6 | C10H12O | 148.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1125-78-6
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00001738
InChI Key UMKXSOXZAXIOPJ-UHFFFAOYSA-N
Synonym 5,6,7,8-tetrahydro-2-naphthol, 6-tetralinol, 6-hydroxytetralin, 2-naphthalenol, 5,6,7,8-tetrahydro, 5,6,7,8-tetrahydro-beta-naphthol, ac-beta-tetralol, 5,6,7,8-tetrahydro-2-hydroxynaphthalene, 2-naphthol, 5,6,7,8-tetrahydro, unii-tmr02i7n8s, 5,6,7,8-tetrahydro-2-naphthalenol
PubChem CID 14305
ChEBI CHEBI:34448
IUPAC Name 5,6,7,8-tetrahydronaphthalen-2-ol
SMILES C1CCC2=C(C1)C=CC(=C2)O

Specifications

Melting Point 60°C to 63°C
Boiling Point 275°C to 276°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1101302
Formula Weight 148.21
Percent Purity 98%
Chemical Name or Material 5,6,7,8-Tetrahydro-2-naphthol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-410-4
RTECSNumber QL5300000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only